Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689140.2 | XP_005689197.1 | 2 - 3 | 459 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |