Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046787.1 | XP_017902276.1 | 3 | 543 | 102 | |
XM_018046788.1 | XP_017902277.1 | 3 | 537 | 102 | ||
XM_018046789.1 | XP_017902278.1 | 3 | 535 | 102 | ||
XM_018046790.1 | XP_017902279.1 | 4 | 2040 | 102 | ||
XM_018046791.1 | XP_017902280.1 | 4 | 636 | 102 | ||
XM_018046792.1 | XP_017902281.1 | 2 | 412 | 102 | ||
XM_018046793.1 | XP_017902282.1 | 2 | 406 | 102 | ||
XM_018046794.1 | XP_017902283.1 | 2 | 350 | 102 | ||
XM_018046795.1 | XP_017902284.1 | 2 | 421 | 102 | ||
GenBank mRNA | JO432365.2 | - | 1 | 1215 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |