Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172948.2 | NP_001166419.1 | 2 - 3 | 194 | 77 | |
GenBank mRNA | AF257212.2 | - | 2 - 3 | 195 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Cavia porcellus Probe spans exons |