Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173045.1 | NP_001166516.1 | 5 - 6 | 802 | 83 | |
GenBank mRNA | FJ234418.1 | - | 5 - 6 | 802 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |