Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CapaR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275246.1 | NP_001262175.1 | 4 - 5 | 836 | 82 | |
NM_206418.3 | NP_996140.1 | 4 - 5 | 836 | 82 | ||
GenBank mRNA | AF505865.1 | - | 4 - 5 | 1018 | 82 | |
AF522193.1 | - | 4 - 5 | 655 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |