Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299424.1 | NP_001286353.1 | 5 - 6 | 3109 | 99 | |
NM_078992.3 | NP_523716.2 | 5 - 6 | 3136 | 99 | ||
NM_165913.2 | NP_725185.1 | 5 - 6 | 2683 | 99 | ||
GenBank mRNA | BT150366.1 | - | 5 - 6 | 3133 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |