Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheB42c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206042.3 | NP_995764.2 | 4 - 5 | 1392 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |