Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zormin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043112.2 | NP_001036577.2 | 16 - 17 | 9234 | 62 | |
NM_001043114.2 | NP_001036579.2 | 17 - 18 | 10917 | 62 | ||
NM_001274386.1 | NP_001261315.1 | 18 - 19 | 10605 | 62 | ||
NM_001274388.1 | NP_001261317.1 | 18 - 19 | 10989 | 62 | ||
NM_206240.2 | NP_995962.2 | 15 - 16 | 8802 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |