Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259741.1 | NP_001246670.1 | 14 - 15 | 6837 | 65 | |
NM_001259742.1 | NP_001246671.1 | 14 - 15 | 7200 | 65 | ||
NM_001259743.1 | NP_001246672.1 | 14 - 15 | 6900 | 65 | ||
NM_001274651.1 | NP_001261580.1 | 14 - 15 | 7185 | 65 | ||
NM_079249.3 | NP_523973.2 | 15 - 16 | 7191 | 65 | ||
NM_145106.1 | NP_659574.1 | 14 - 15 | 6638 | 65 | ||
NM_176300.1 | NP_788478.1 | 15 - 16 | 6704 | 65 | ||
NM_206303.2 | NP_996025.1 | 15 - 16 | 7191 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster Inventoried |