Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299095.1 | NP_001286024.1 | 7 - 8 | 1328 | 62 | |
NM_136017.3 | NP_609861.1 | 7 - 8 | 1328 | 62 | ||
GenBank mRNA | BT004502.1 | - | 7 - 8 | 1301 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |