Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058010.4 | NP_477358.1 | 2 - 3 | 361 | 74 | |
NM_166102.2 | NP_725476.1 | 2 - 3 | 361 | 74 | ||
GenBank mRNA | AF044734.1 | - | 2 - 3 | 127 | 74 | |
AY058794.1 | - | 2 - 3 | 336 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |