Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202020.1 | NP_001188949.1 | 3 - 4 | 1177 | 78 | |
NM_001202026.2 | NP_001188955.2 | 17 - 18 | 21392 | 78 | ||
NM_001274089.1 | NP_001261018.1 | 17 - 18 | 21392 | 78 | ||
NM_001274090.1 | NP_001261019.1 | 17 - 18 | 21760 | 78 | ||
NM_166129.3 | NP_725510.2 | 17 - 18 | 21760 | 78 | ||
GenBank mRNA | AY113646.1 | - | 3 - 4 | 1178 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |