Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079032.4 | NP_523756.1 | 13 - 14 | 3520 | 68 | |
NM_166138.5 | NP_725523.3 | 12 - 13 | 3271 | 68 | ||
NM_176183.3 | NP_788363.1 | 14 - 15 | 4315 | 68 | ||
NM_176184.4 | NP_788364.2 | 13 - 14 | 4066 | 68 | ||
GenBank mRNA | AF461100.1 | - | 12 - 13 | 3418 | 68 | |
AF461101.1 | - | 12 - 13 | 2872 | 68 | ||
AY069076.1 | - | 13 - 14 | 3493 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |