Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trf4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103443.2 | NP_001096913.1 | 4 - 5 | 1984 | 77 | |
NM_001103444.2 | NP_001096914.1 | 4 - 5 | 1971 | 77 | ||
NM_001272411.1 | NP_001259340.1 | 3 - 4 | 2276 | 77 | ||
NM_132262.3 | NP_572490.2 | 3 - 4 | 2241 | 77 | ||
NM_206658.4 | NP_996381.1 | 3 - 4 | 2403 | 77 | ||
NM_206659.3 | NP_996382.2 | 3 - 4 | 2208 | 77 | ||
NM_206660.2 | NP_996383.2 | 3 - 4 | 1978 | 77 | ||
GenBank mRNA | BT010282.1 | - | 3 - 4 | 2090 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() |