Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014733.5 | NP_001014733.1 | 5 - 6 | 1044 | 78 | |
NM_001014734.4 | NP_001014734.1 | 5 - 6 | 1044 | 78 | ||
NM_001258707.2 | NP_001245636.1 | 5 - 6 | 1044 | 78 | ||
NM_001258708.2 | NP_001245637.1 | 5 - 6 | 1044 | 78 | ||
NM_001258709.2 | NP_001245638.1 | 4 - 5 | 971 | 78 | ||
NM_001258710.2 | NP_001245639.1 | 5 - 6 | 1044 | 78 | ||
NM_001272548.1 | NP_001259477.1 | 4 - 5 | 974 | 78 | ||
NM_001370017.1 | NP_001356943.1 | 5 - 6 | 1044 | 78 | ||
NM_001370018.1 | NP_001356944.1 | 5 - 6 | 1044 | 78 | ||
NM_001370019.1 | NP_001356945.1 | 5 - 6 | 1044 | 78 | ||
NM_078578.5 | NP_511133.2 | 5 - 6 | 1044 | 78 | ||
NM_206694.4 | NP_996417.1 | 5 - 6 | 1054 | 78 | ||
NM_206696.6 | NP_996419.1 | 5 - 6 | 1044 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |