Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258868.3 | NP_001245797.1 | 5 - 6 | 1526 | 58 | |
NM_001258869.3 | NP_001245798.1 | 5 - 6 | 1409 | 58 | ||
NM_001298597.1 | NP_001285526.1 | 5 - 6 | 1381 | 58 | ||
NM_134617.5 | NP_608461.1 | 4 - 5 | 1595 | 58 | ||
GenBank mRNA | BT099938.1 | - | 4 - 5 | 1581 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |