Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272915.1 | NP_001259844.1 | 1 | 927 | 66 | |
NM_134702.3 | NP_608546.1 | 1 | 927 | 66 | ||
GenBank mRNA | BT023302.1 | - | 1 | 897 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |