Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iPLA2-VIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140109.2 | NP_648366.2 | 4 - 5 | 1576 | 99 | |
NM_168383.2 | NP_729565.2 | 5 - 6 | 1714 | 99 | ||
NM_168384.2 | NP_729566.2 | 4 - 5 | 1992 | 99 | ||
NM_168385.2 | NP_729567.2 | 5 - 6 | 1899 | 99 | ||
GenBank mRNA | BT125951.1 | - | 4 - 5 | 1453 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |