Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk29A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103650.2 | NP_001097120.1 | 8 - 9 | 3860 | 92 | |
NM_001103651.3 | NP_001097121.1 | 12 - 13 | 2521 | 92 | ||
NM_001273314.1 | NP_001260243.1 | 8 - 9 | 2170 | 92 | ||
NM_001273315.1 | NP_001260244.1 | 11 - 12 | 2694 | 92 | ||
NM_057397.6 | NP_476745.1 | 12 - 13 | 2854 | 92 | ||
NM_057398.4 | NP_476746.1 | 9 - 10 | 2144 | 92 | ||
NM_164804.2 | NP_723369.1 | 12 - 13 | 2542 | 92 | ||
NM_164805.2 | NP_723370.1 | 9 - 10 | 2147 | 92 | ||
GenBank mRNA | AY069457.1 | - | 8 - 9 | 3991 | 92 | |
AY128441.1 | - | 12 - 13 | 2809 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |