Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135024.2 | NP_608868.1 | 1 - 2 | 698 | 92 | |
GenBank mRNA | BT150242.1 | - | 1 - 2 | 643 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |