Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135068.4 | NP_608912.1 | 1 - 2 | 237 | 72 | |
GenBank mRNA | AY069176.1 | - | 1 - 2 | 218 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |