Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG7542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300170.1 | NP_001287099.1 | 1 | 765 | 71 | |
NM_140738.2 | NP_648995.3 | 1 | 765 | 71 | ||
GenBank mRNA | AY089278.1 | - | 1 | 803 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |