Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169519.2 | NP_001162990.1 | 10 - 11 | 2290 | 79 | |
NM_001169520.3 | NP_001162991.1 | 10 - 11 | 2452 | 79 | ||
NM_001259120.1 | NP_001246049.1 | 10 - 11 | 2445 | 79 | ||
NM_001316398.1 | NP_001303327.1 | 10 - 11 | 2445 | 79 | ||
NM_078863.6 | NP_523587.4 | 10 - 11 | 2290 | 79 | ||
NM_165182.3 | NP_724000.1 | 10 - 11 | 2290 | 79 | ||
NM_165183.3 | NP_724001.1 | 10 - 11 | 2290 | 79 | ||
NM_165184.3 | NP_724002.2 | 10 - 11 | 2432 | 79 | ||
NM_165187.3 | NP_724005.1 | 10 - 11 | 2290 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |