Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plc21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272889.1 | NP_001259818.1 | 15 - 16 | 4257 | 68 | |
NM_057503.5 | NP_476851.2 | 16 - 17 | 4394 | 68 | ||
NM_057504.5 | NP_476852.1 | 16 - 17 | 4443 | 68 | ||
NM_205882.2 | NP_995604.2 | 16 - 17 | 4376 | 68 | ||
NM_205883.2 | NP_995605.1 | 16 - 17 | 4412 | 68 | ||
NM_205884.3 | NP_995606.1 | 16 - 17 | 4440 | 68 | ||
GenBank mRNA | AY051657.1 | - | 16 - 17 | 4403 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |