Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FucTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298850.1 | NP_001285779.1 | 1 - 2 | 355 | 79 | |
NM_135444.5 | NP_609288.4 | 1 - 2 | 355 | 79 | ||
GenBank mRNA | AY061932.1 | - | 1 - 2 | 233 | 79 | |
BT023506.1 | - | 1 - 2 | 332 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |