Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135172.2 | NP_609016.2 | 1 - 2 | 980 | 119 | |
GenBank mRNA | AY226540.1 | - | 1 - 2 | 980 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |