Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298746.1 | NP_001285675.1 | 3 - 4 | 4638 | 91 | |
NM_135191.3 | NP_609035.2 | 3 - 4 | 4638 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |