Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135313.3 | NP_609157.3 | 2 - 3 | 380 | 54 | |
GenBank mRNA | AY070616.1 | - | 2 - 3 | 377 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |