Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RfC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135555.3 | NP_609399.1 | 3 - 4 | 629 | 64 | |
GenBank mRNA | AF247499.1 | - | 3 - 4 | 617 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |