Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165250.4 | NP_724107.1 | 7 - 8 | 2141 | 98 | |
GenBank mRNA | M27813.1 | - | 7 - 8 | 2139 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |