Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273627.1 | NP_001260556.1 | 1 - 2 | 1700 | 103 | |
NM_136091.4 | NP_609935.1 | 2 - 3 | 1615 | 103 | ||
NM_165273.2 | NP_724153.1 | 1 - 2 | 1700 | 103 | ||
GenBank mRNA | AY069519.1 | - | 1 - 2 | 1661 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |