Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259184.1 | NP_001246113.1 | 4 - 5 | 2958 | 59 | |
NM_001273711.1 | NP_001260640.1 | 4 - 5 | 3320 | 59 | ||
NM_057633.5 | NP_476981.1 | 4 - 5 | 3320 | 59 | ||
NM_165341.3 | NP_724285.1 | 4 - 5 | 3042 | 59 | ||
GenBank mRNA | BT021398.1 | - | 4 - 5 | 3298 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |