Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Eh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078910.4 | NP_523634.1 | 2 - 3 | 351 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |