Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042810.3 | NP_001036275.1 | 14 - 15 | 2593 | 73 | |
NM_001201663.2 | NP_001188592.1 | 14 - 15 | 2587 | 73 | ||
NM_001272629.1 | NP_001259558.1 | 13 - 14 | 2542 | 73 | ||
NM_078603.4 | NP_511158.2 | 14 - 15 | 2593 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |