Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103531.2 | NP_001097001.1 | 4 - 5 | 712 | 71 | |
NM_001272692.1 | NP_001259621.1 | 4 - 5 | 949 | 71 | ||
NM_078648.4 | NP_523372.2 | 3 - 4 | 951 | 71 | ||
GenBank mRNA | AY061256.1 | - | 3 - 4 | 951 | 71 | |
BT126051.1 | - | 4 - 5 | 692 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |