Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: if | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169306.2 | NP_001162777.1 | 11 - 12 | 4323 | 93 | |
NM_001169307.1 | NP_001162778.1 | 11 - 12 | 4458 | 93 | ||
NM_078654.2 | NP_523378.2 | 11 - 12 | 4364 | 93 | ||
NM_167544.2 | NP_728021.2 | 10 - 11 | 4289 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |