Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274099.1 | NP_001261028.1 | 2 - 3 | 634 | 90 | |
NM_001274100.1 | NP_001261029.1 | 3 - 4 | 779 | 90 | ||
NM_001274101.1 | NP_001261030.1 | 3 - 4 | 411 | 90 | ||
NM_001274102.1 | NP_001261031.1 | 3 - 4 | 411 | 90 | ||
NM_001274103.1 | NP_001261032.1 | 3 - 4 | 779 | 90 | ||
NM_166186.3 | NP_725599.2 | 3 - 4 | 411 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |