Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274099.1 | NP_001261028.1 | 6 - 7 | 1379 | 87 | |
NM_001274101.1 | NP_001261030.1 | 7 - 8 | 1156 | 87 | ||
NM_001274102.1 | NP_001261031.1 | 8 - 9 | 1195 | 87 | ||
NM_166186.3 | NP_725599.2 | 8 - 9 | 1195 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |