Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057616.6 | NP_476964.1 | 2 - 3 | 228 | 85 | |
NM_166383.4 | NP_725943.1 | 3 - 4 | 251 | 85 | ||
GenBank mRNA | BT024929.1 | - | 2 - 3 | 222 | 85 | |
L22959.1 | - | 1 - 2 | 190 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |