Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clumsy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078886.3 | NP_523610.2 | 9 - 10 | 2426 | 77 | |
GenBank mRNA | BT150351.1 | - | 9 - 10 | 2399 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |