Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rho-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299395.1 | NP_001286324.1 | 4 - 5 | 888 | 72 | |
NM_078980.4 | NP_523704.1 | 4 - 5 | 888 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |