Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299885.1 | NP_001286814.1 | 7 - 8 | 1974 | 73 | |
NM_057365.4 | NP_476713.1 | 6 - 7 | 1493 | 73 | ||
NM_166653.2 | NP_726408.1 | 6 - 7 | 1529 | 73 | ||
GenBank mRNA | AY058350.1 | - | 6 - 7 | 1479 | 73 | |
U36925.1 | - | 6 - 7 | 1464 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |