Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cals | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272157.1 | NP_001259086.1 | 6 - 7 | 2107 | 72 | |
NM_001297793.1 | NP_001284722.1 | 6 - 7 | 2110 | 72 | ||
NM_079893.3 | NP_524632.1 | 6 - 7 | 2107 | 72 | ||
NM_166823.2 | NP_726646.1 | 6 - 7 | 2110 | 72 | ||
GenBank mRNA | AJ289018.1 | - | 6 - 7 | 2073 | 72 | |
AY121644.1 | - | 6 - 7 | 2075 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |