Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169695.2 | NP_001163166.1 | 2 - 3 | 332 | 60 | |
NM_001299547.1 | NP_001286476.1 | 2 - 3 | 320 | 60 | ||
NM_079034.5 | NP_523758.3 | 2 - 3 | 379 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |