Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274097.2 | NP_001261026.1 | 6 - 7 | 1347 | 88 | |
NM_079039.4 | NP_523763.2 | 6 - 7 | 1347 | 88 | ||
GenBank mRNA | AF439752.1 | - | 5 - 6 | 1080 | 88 | |
AY051579.1 | - | 6 - 7 | 1367 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster Inventoried |