Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079054.5 | NP_523778.2 | 7 - 8 | 2168 | 88 | |
GenBank mRNA | BT128787.1 | - | 7 - 8 | 2152 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |