Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169855.2 | NP_001163326.1 | 1 - 2 | 137 | 77 | |
NM_079163.4 | NP_523887.2 | 1 - 2 | 137 | 77 | ||
NM_167927.3 | NP_728695.1 | 1 - 2 | 137 | 77 | ||
GenBank mRNA | AY061475.1 | - | 1 - 2 | 175 | 77 | |
BT030128.1 | - | 1 - 2 | 116 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |