Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043115.3 | NP_001036580.1 | 6 - 7 | 3106 | 59 | |
NM_001202108.2 | NP_001189037.1 | 7 - 8 | 3196 | 59 | ||
NM_001202109.2 | NP_001189038.1 | 6 - 7 | 2986 | 59 | ||
NM_001202110.2 | NP_001189039.1 | 6 - 7 | 3061 | 59 | ||
NM_001259655.2 | NP_001246584.1 | 6 - 7 | 3061 | 59 | ||
NM_001259656.2 | NP_001246585.1 | 6 - 7 | 3061 | 59 | ||
NM_001274416.1 | NP_001261345.1 | 6 - 7 | 3091 | 59 | ||
NM_079170.6 | NP_523894.2 | 7 - 8 | 3094 | 59 | ||
NM_167967.4 | NP_728783.1 | 6 - 7 | 3106 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |