Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K68D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169949.2 | NP_001163420.1 | 5 - 6 | 1961 | 83 | |
NM_079304.4 | NP_524028.2 | 4 - 5 | 2310 | 83 | ||
NM_168475.3 | NP_729743.1 | 5 - 6 | 2009 | 83 | ||
GenBank mRNA | AY058587.1 | - | 4 - 5 | 2252 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |