Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079320.3 | NP_524044.1 | 1 - 2 | 176 | 61 | |
GenBank mRNA | BT004490.1 | - | 1 - 2 | 147 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |